Assay Details
| Gene Symbol: H60c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014573.1 | - | 2 - 3 | 392 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA B020014A21 gene | B020014A21Rik | -123.0kb | Sense | Chr. 10 - 3125091 - 3133193 on NCBI Build GRCm38 | |
| Downstream |
protein phosphatase 1, regulatory (inhibitor) subunit 14c |
Ppp1r14c |
6330514J04Rik, KEPI |
106.1kb | Anti-sense | Chr. 10 - 3366071 - 3464979 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

H60c
Genomic Map