Assay Details
Gene Symbol: Vmn1r66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134230.3 | NP_598991.2 | 1 | 1424 | 61 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203277 | BC109342.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203277 | AF394952.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203277 | BC109341.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203277 | NM_134230.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203277 | BC109342.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203277 | AF394952.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203277 | BC109341.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P203277 | NM_134230.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78546 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159728 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203277 | BC109342.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203277 | AF394952.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203277 | BC109341.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203277 | NM_134230.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203277 | BC109342.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203277 | AF394952.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203277 | BC109341.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P203277 | NM_134230.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203277 | BC109342.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203277 | AF394952.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203277 | BC109341.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203277 | NM_134230.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203277 | BC109342.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203277 | AF394952.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203277 | BC109341.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P203277 | NM_134230.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965528 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965534 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965549 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965541 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965548 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965557 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965566 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965555 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965544 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965551 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965533 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965543 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965561 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965531 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965559 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965526 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965545 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965525 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965518 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965538 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965536 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965556 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965530 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965560 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965524 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965520 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965539 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965521 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965527 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965563 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965564 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965514 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965513 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965565 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965542 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965552 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24965523 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159728 | - | - |
Whole Mouse Genome G4122A | A_51_P203277 | BC109342.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P203277 | AF394952.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P203277 | BC109341.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P203277 | NM_134230.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P203277 | BC109342.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P203277 | AF394952.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P203277 | BC109341.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P203277 | NM_134230.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126174 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159728 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06511613 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06511612 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06449613 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06449618 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06304934 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06511622 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06449622 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06449621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24965570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06304931 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965561 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965514 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06595157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06205855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06595159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965520 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06304928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965574 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06386481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965523 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24965543 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10559948 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559951 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559947 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559950 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559949 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10559946 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700020704.mm.1 | TC0700002308.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700020704.mm.1 | NM_134230.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700020704.mm.1 | NM_134230.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 514302 | - | - |
ABI Mouse Genome Survey Microarray | 757892 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3840070 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003840070 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2443381 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2443381 | - | - |
mouse-6 v1.1 (Illumina) | scl31762.1.248_30-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3840070 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl31762.1.248_30-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840070 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2443381 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2443381 | - | - |
