Assay Details
| Gene Symbol: Zmpste24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172700.2 | NP_766288.1 | 2 - 3 | 503 | 70 | |
| GenBank mRNA | AJ487544.1 | - | 2 - 3 | 469 | 70 | |
| AK083566.1 | - | 2 - 3 | 503 | 70 | ||
| AK170946.1 | - | 2 - 3 | 461 | 70 | ||
| AY029194.1 | - | 2 - 3 | 259 | 70 | ||
| BC023740.1 | - | 2 - 3 | 445 | 70 | ||
| BC138578.1 | - | 2 - 3 | 371 | 8861598 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zmpste24
Genomic Map