Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400010225.mm.1 |
NM_001159630.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010225.mm.1 |
NM_001159630.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010225.mm.1 |
NM_013883.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010225.mm.1 |
NM_013883.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010225.mm.1 |
TC0400001347.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010226.mm.1 |
NM_001159630.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010226.mm.1 |
NM_001159630.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010226.mm.1 |
NM_013883.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010226.mm.1 |
NM_013883.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010226.mm.1 |
TC0400001347.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010228.mm.1 |
NM_001159630.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010228.mm.1 |
NM_001159630.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010228.mm.1 |
NM_013883.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010228.mm.1 |
NM_013883.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010228.mm.1 |
TC0400001347.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010229.mm.1 |
NM_001159630.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010229.mm.1 |
NM_001159630.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010229.mm.1 |
NM_013883.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010229.mm.1 |
NM_013883.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010229.mm.1 |
TC0400001347.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010230.mm.1 |
NM_001159630.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010230.mm.1 |
NM_001159630.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010230.mm.1 |
NM_013883.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010230.mm.1 |
NM_013883.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010230.mm.1 |
TC0400001347.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010234.mm.1 |
NM_001159630.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010234.mm.1 |
NM_001159630.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010234.mm.1 |
NM_013883.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010234.mm.1 |
NM_013883.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010234.mm.1 |
TC0400001347.mm.1 |
MTA TC |