Assay Details
| Gene Symbol: Smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133716.3 | NP_598477.2 | 1 - 2 | 503 | 85 | |
| GenBank mRNA | AK145042.1 | - | 1 - 2 | 488 | 85 | |
| AK152757.1 | - | 1 - 2 | 470 | 85 | ||
| BC031581.1 | - | 1 - 2 | 492 | 85 | ||
| BC032217.1 | - | 1 - 2 | 479 | 85 | ||
| BC052413.1 | - | 1 - 2 | 503 | 5720177 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smap2
Genomic Map