Assay Details
| Gene Symbol: Smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133716.3 | NP_598477.2 | 8 - 9 | 1237 | 112 | |
| GenBank mRNA | BC031581.1 | - | 8 - 9 | 1226 | 112 | |
| BC032217.1 | - | 8 - 9 | 1213 | 112 | ||
| BC052413.1 | - | 8 - 9 | 1237 | 5720177 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smap2
Genomic Map