Assay Details
| Gene Symbol: Smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133716.3 | NP_598477.2 | 9 - 10 | 1560 | 71 | |
| GenBank mRNA | AK007666.1 | - | 1 - 2 | 161 | 71 | |
| BC031581.1 | - | 9 - 10 | 1549 | 71 | ||
| BC032217.1 | - | 9 - 10 | 1536 | 71 | ||
| BC052413.1 | - | 9 - 10 | 1560 | 5720177 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007666.1 transcripts.
Set Membership: |
|

Smap2
Genomic Map