Assay Details
| Gene Symbol: Aim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172393.2 | NP_765981.2 | 12 - 13 | 4902 | 143 | |
| GenBank mRNA | AF397913.1 | - | 6 - 7 | 609 | 143 | |
| AK041082.1 | - | 11 - 12 | 1518 | 143 | ||
| AK088102.1 | - | 11 - 12 | 1574 | 143 | ||
| BC035551.1 | - | 11 - 12 | 2054 | 143 | ||
| BC059272.1 | - | 11 - 12 | 2059 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aim1
Genomic Map