Assay Details
| Gene Symbol: Aim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172393.2 | NP_765981.2 | 4 - 5 | 3884 | 75 | |
| GenBank mRNA | AK041082.1 | - | 3 - 4 | 500 | 75 | |
| AK088102.1 | - | 3 - 4 | 556 | 75 | ||
| BC035551.1 | - | 3 - 4 | 1036 | 75 | ||
| BC059272.1 | - | 3 - 4 | 1041 | 75 | ||
More Information
Set Membership: |
|

Aim1
Genomic Map