Assay Details
| Gene Symbol: Hcrtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163027.1 | NP_001156499.1 | 7 - 8 | 1258 | 88 | |
| NM_198959.2 | NP_945197.2 | 7 - 8 | 1284 | 88 | ||
| GenBank mRNA | AF394596.1 | - | 3 - 4 | 574 | 88 | |
| AK140137.1 | - | 7 - 8 | 1258 | 88 | ||
| AY336083.1 | - | 7 - 8 | 1284 | 88 | ||
| BC119582.2 | - | 7 - 8 | 1156 | 88 | ||
| BC119583.2 | - | 6 - 7 | 1140 | 40058834 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hcrtr1
Genomic Map