Assay Details
| Gene Symbol: Khdrbs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011317.4 | NP_035447.3 | 6 - 7 | 1251 | 69 | |
| NR_045036.1 | 6 - 7 | 1251 | 69 | |||
| GenBank mRNA | AK044927.1 | - | 6 - 7 | 1248 | 69 | |
| AK050520.1 | - | 6 - 7 | 1246 | 69 | ||
| AK146815.1 | - | 6 - 7 | 1213 | 69 | ||
| AK150687.1 | - | 6 - 7 | 1230 | 69 | ||
| AK152084.1 | - | 6 - 7 | 1230 | 69 | ||
| AK204780.1 | - | 1 - 2 | 174 | 69 | ||
| BC002051.1 | - | 6 - 7 | 1207 | 3490667 | 69 | |
| U17046.1 | - | 6 - 7 | 1208 | 69 | ||
| U17961.1 | - | 6 - 7 | 1219 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Khdrbs1
Genomic Map