Assay Details
| Gene Symbol: Il23r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144548.1 | NP_653131.2 | 9 - 10 | 1317 | 98 | |
| GenBank mRNA | AF461423.1 | - | 9 - 10 | 1317 | 98 | |
| BC112425.1 | - | 9 - 10 | 1300 | 40062206 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il23r
Genomic Map