Assay Details
| Gene Symbol: E230015B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042791.1 | - | 3 - 4 | 368 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Riken cDNA 1810020O05 gene | 1810020O05Rik | Anti-sense | Chr. 6 - 87675593 - 87690847 on NCBI Build GRCm38 | ||
| Downstream | EF hand and coiled-coil domain containing 1 | Efcc1 | Ccdc48 | 54.9kb | Sense | Chr. 6 - 87730820 - 87755912 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

E230015B07Rik
Genomic Map