Assay Details
| Gene Symbol: 4930511E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040761.1 | 6 - 7 | 1200 | 156 | ||
| GenBank mRNA | AK019674.1 | - | 6 - 7 | 1202 | 156 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine-rich repeats and immunoglobulin-like domains 1 | Lrig1 |
D6Bwg0781e, Img, LIG-1 |
-248.7kb | Anti-sense | Chr. 6 - 94604529 - 94700253 on NCBI Build GRCm38 |
| Downstream | kelch repeat and BTB (POZ) domain containing 8 | Kbtbd8 |
AU040152, SSEC-51, SSEC51, Takrp, mKIAA1842 |
167.8kb | Sense | Chr. 6 - 95117880 - 95129793 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930511E03Rik
Genomic Map