Assay Details
| Gene Symbol: Nlrp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172481.2 | NP_766069.1 | 5 - 6 | 2158 | 128 | |
| GenBank mRNA | AK054426.1 | - | 5 - 6 | 2159 | 128 | |
| AK162217.1 | - | 2 - 3 | 890 | 128 | ||
| AY355342.1 | - | 5 - 6 | 2159 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp4b
Genomic Map