Assay Details
| Gene Symbol: Rps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009098.2 | NP_033124.1 | 1 - 2 | 73 | 68 | |
| GenBank mRNA | AK010650.1 | - | 1 - 2 | 73 | 68 | |
| AK011048.1 | - | 1 - 2 | 42 | 68 | ||
| AK012665.1 | - | 1 - 2 | 45 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010650.1, AK011048.1, AK012665.1, NM_009098.2 transcripts.
Set Membership: |
|

Rps8
Genomic Map