Assay Details
| Gene Symbol: Ptch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008958.2 | NP_032984.1 | 13 - 14 | 1711 | 84 | |
| GenBank mRNA | AB010833.1 | - | 13 - 14 | 1711 | 84 | |
| BC058397.1 | - | 13 - 14 | 1966 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptch2
Genomic Map