Assay Details
| Gene Symbol: Ptch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008958.2 | NP_032984.1 | 4 - 5 | 525 | 98 | |
| GenBank mRNA | BC058397.1 | - | 4 - 5 | 785 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptch2
Genomic Map