Assay Details
| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013807.2 | NP_038835.2 | 7 - 8 | 1040 | 99 | |
| GenBank mRNA | AK156754.1 | - | 7 - 8 | 1052 | 99 | |
| BC031180.1 | - | 7 - 8 | 1032 | 4988360 | 99 | |
| BC063051.1 | - | 7 - 8 | 1040 | 5718130 | 99 | |
| U21392.1 | - | 7 - 8 | 887 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk3
Genomic Map