Assay Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009478.3 | NP_033504.2 | 9 - 10 | 972 | 72 | |
GenBank mRNA | AK002844.1 | - | 9 - 10 | 961 | 72 | |
AK007470.1 | - | 9 - 10 | 963 | 72 | ||
AK172426.1 | - | 9 - 10 | 983 | 72 | ||
BC008109.1 | - | 9 - 10 | 972 | 3481699 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | AK172426.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | BC008109.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | AK002844.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | AK172426.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | BC008109.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P282092 | AK002844.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856947 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475495 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162692 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | AK172426.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | BC008109.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | AK002844.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | AK172426.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | BC008109.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P282092 | AK002844.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | AK172426.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | BC008109.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | AK002844.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | AK172426.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | BC008109.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P282092 | AK002844.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231904 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231870 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231891 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231873 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231883 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231895 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231887 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23231884 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162692 | - | - |
Whole Mouse Genome G4122A | A_51_P282092 | AK172426.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P282092 | BC008109.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P282092 | AK002844.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P282092 | AK172426.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P282092 | BC008109.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P282092 | NM_009478.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P282092 | AK002844.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134060 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162692 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798082 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798089 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798084 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798073 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798100 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798088 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03798096 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03219573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23231876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231903 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03261655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03798082 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04037944 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231874 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03755530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04037950 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23231887 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10515391 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515392 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515387 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515394 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515386 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515393 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515388 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515395 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515389 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515390 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443849_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417206_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10515385 | - | - |
Mouse Genome 430 2.0 Array | 1443849_x_at | - | - |
Mouse Genome 430 2.0 Array | 1417206_at | - | - |
Mouse Genome 430 2.0 Array | 1443849_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1417206_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22275_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22275_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400026824.mm.1 | TC0400003357.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400026824.mm.1 | NM_009478.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400026823.mm.1 | TC0400003357.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400026823.mm.1 | NM_009478.4 | Refseq |
Mouse Expression Set 430 | 1417206_PM_at | - | - |
Mouse Expression Set 430 | 1417206_at | - | - |
Mouse Expression Set 430 | 1443849_PM_x_at | - | - |
Mouse Expression Set 430 | 1443849_x_at | - | - |
Murine Genome U74v2 Set | 161328_i_at | - | - |
Murine Genome U74v2 Set | 161328_PM_i_at | - | - |
Murine Genome U74v2 Set | 94275_at | - | - |
Murine Genome U74v2 Set | 94275_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417206_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417206_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417206_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417206_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443849_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443849_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 672607 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2470494 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002470494 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2453695 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2453695 | - | - |
mouse-6 v1.1 (Illumina) | scl23946.8.1_41-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2470494 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl23946.8.1_41-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470494 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2453695 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2453695 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl23946.8.1_41-S | - | - |
