Assay Details
| Gene Symbol: Phactr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175306.4 | NP_780515.2 | 3 - 4 | 327 | 113 | |
| GenBank mRNA | AK049209.1 | - | 3 - 4 | 290 | 113 | |
| BC075672.1 | - | 3 - 4 | 289 | 30549496 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phactr4
Genomic Map