Assay Details
| Gene Symbol: Chmp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026885.3 | NP_081161.1 | 3 - 4 | 539 | 74 | |
| GenBank mRNA | AK005267.1 | - | 3 - 4 | 507 | 74 | |
| BC012230.1 | - | 3 - 4 | 480 | 4024872 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chmp2a
Genomic Map