Assay Details
| Gene Symbol: Smpdl3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133888.2 | NP_598649.1 | 3 - 4 | 499 | 71 | |
| GenBank mRNA | AK136602.1 | - | 2 - 3 | 161 | 71 | |
| AK154599.1 | - | 3 - 4 | 525 | 71 | ||
| AK166253.1 | - | 3 - 4 | 475 | 71 | ||
| BC009087.1 | - | 3 - 4 | 499 | 2810972 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpdl3b
Genomic Map