Assay Details
| Gene Symbol: Taf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025579.3 | NP_079855.2 | 3 - 4 | 443 | 76 | |
| GenBank mRNA | AK007811.1 | - | 3 - 4 | 406 | 76 | |
| AK132927.1 | - | 3 - 4 | 444 | 76 | ||
| AK139748.1 | - | 3 - 4 | 357 | 76 | ||
| BC019668.1 | - | 3 - 4 | 388 | 3493047 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf12
Genomic Map