Assay Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197082.1 | NP_001184011.1 | 6 - 7 | 690 | 69 | |
| NM_133878.3 | NP_598639.1 | 5 - 6 | 651 | 69 | ||
| GenBank mRNA | AK076089.1 | - | 5 - 6 | 652 | 69 | |
| AK150153.1 | - | 5 - 6 | 603 | 69 | ||
| AK150610.1 | - | 5 - 6 | 601 | 69 | ||
| AK153193.1 | - | 5 - 6 | 601 | 69 | ||
| AK155225.1 | - | 3 - 4 | 233 | 69 | ||
| BC019807.1 | - | 5 - 6 | 563 | 4456795 | 69 | |
| BC057645.1 | - | 6 - 7 | 645 | 3591859 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcc1
Genomic Map