Assay Details
| Gene Symbol: Epb4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128606.1 | NP_001122078.1 | 5 - 6 | 1045 | 99 | |
| NM_001128607.1 | NP_001122079.1 | 5 - 6 | 955 | 99 | ||
| NM_183428.3 | NP_906273.3 | 5 - 6 | 2307 | 99 | ||
| GenBank mRNA | AB370261.1 | - | 4 - 5 | 830 | 99 | |
| AK220462.1 | - | 5 - 6 | 952 | 99 | ||
| BC068138.1 | - | 5 - 6 | 915 | 99 | ||
| BC079875.1 | - | 5 - 6 | 874 | 6844082 | 99 | |
| CN537562.1 | - | 3 - 4 | 363 | 99 | ||
| L00919.1 | - | 6 - 7 | 2382 | 99 | ||
| LK021332.1 | - | 3 - 4 | 200 | 99 | ||
| LK021333.1 | - | 3 - 4 | 200 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epb4.1
Genomic Map