Assay Details
| Gene Symbol: Sec14l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029937.2 | NP_001025108.1 | 5 - 6 | 523 | 92 | |
| GenBank mRNA | BC138982.1 | - | 5 - 6 | 523 | 8862004 | 92 |
| BC138985.1 | - | 5 - 6 | 523 | 8862007 | 92 | |
| BC145485.1 | - | 5 - 6 | 566 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec14l3
Genomic Map