Assay Details
| Gene Symbol: Eif4enif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166547.1 | NP_001160019.1 | 14 - 15 | 2212 | 67 | |
| NM_001166548.1 | NP_001160020.1 | 13 - 14 | 2144 | 67 | ||
| NM_001166549.1 | NP_001160021.1 | 13 - 14 | 2248 | 67 | ||
| NM_023743.2 | NP_076232.2 | 14 - 15 | 2227 | 67 | ||
| GenBank mRNA | AB031388.1 | - | 14 - 15 | 2048 | 67 | |
| AK012082.1 | - | 4 - 5 | 631 | 67 | ||
| AK041598.1 | - | 13 - 14 | 2106 | 67 | ||
| AK088425.1 | - | 9 - 10 | 1314 | 67 | ||
| AK128990.1 | - | 13 - 14 | 2246 | 67 | ||
| AK165678.1 | - | 14 - 15 | 2214 | 67 | ||
| BC033410.1 | - | 14 - 15 | 2062 | 5359290 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif4enif1
Genomic Map