Assay Details
| Gene Symbol: Limk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034030.2 | NP_001029202.1 | 7 - 8 | 857 | 69 | |
| NM_010718.4 | NP_034848.1 | 11 - 12 | 1496 | 69 | ||
| NM_173053.1 | NP_774958.1 | 10 - 11 | 1627 | 69 | ||
| GenBank mRNA | AB005131.1 | - | 11 - 12 | 1349 | 69 | |
| AB005132.1 | - | 10 - 11 | 1611 | 69 | ||
| AB008117.1 | - | 11 - 12 | 1432 | 69 | ||
| AB012092.1 | - | 7 - 8 | 857 | 69 | ||
| AB012291.1 | - | 7 - 8 | 857 | 69 | ||
| AK034900.1 | - | 2 - 3 | 158 | 69 | ||
| AK146099.1 | - | 10 - 11 | 1584 | 69 | ||
| BC007129.1 | - | 11 - 12 | 1439 | 3496836 | 69 | |
| BC086629.1 | - | 10 - 11 | 1557 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Limk2
Genomic Map