Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100020648.mm.1 |
NM_001190406.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020648.mm.1 |
NM_001190406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020648.mm.1 |
NM_030228.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020648.mm.1 |
NM_030228.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020648.mm.1 |
TC1100002124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020647.mm.1 |
NM_001190406.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020647.mm.1 |
NM_001190406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020647.mm.1 |
NM_030228.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020647.mm.1 |
NM_030228.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020647.mm.1 |
TC1100002124.mm.1 |
MTA TC |