Assay Details
| Gene Symbol: Mrps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026080.2 | NP_080356.1 | 2 - 3 | 149 | 173 | |
| GenBank mRNA | AK003117.1 | - | 2 - 3 | 133 | 173 | |
| AK007338.1 | - | 2 - 3 | 133 | 173 | ||
| AK010590.1 | - | 2 - 3 | 130 | 173 | ||
| AK012461.1 | - | 2 - 3 | 138 | 173 | ||
| AK014099.1 | - | 2 - 3 | 136 | 173 | ||
| BY100364.1 | - | 2 - 3 | 152 | 173 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003117.1, AK007338.1, AK010590.1, AK014099.1, NM_026080.2 transcripts.
Set Membership: |
|

Mrps24
Genomic Map