Assay Details
| Gene Symbol: Gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010292.5 | NP_034422.2 | 1 - 2 | 528 | 62 | |
| GenBank mRNA | L38990.1 | - | 1 - 2 | 46 | 62 | |
| M58755.1 | - | 1 - 2 | 524 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gck
Genomic Map