Assay Details
| Gene Symbol: Urgcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077661.1 | NP_001071129.1 | 1 - 2 | 495 | 69 | |
| GenBank mRNA | AK035580.1 | - | 1 - 2 | 491 | 69 | |
| BC066994.1 | - | 1 - 2 | 376 | 5696000 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035580.1, BC066994.1, NM_001077661.1 transcripts.
Set Membership: |
|

Urgcp
Genomic Map