Assay Details
| Gene Symbol: Upp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159401.1 | NP_001152873.1 | 1 - 2 | 159 | 59 | |
| GenBank mRNA | AL118479.1 | - | 1 - 2 | 159 | 59 | |
| AV447018.1 | - | 1 - 2 | 210 | 59 | ||
| BC036559.1 | - | 1 - 2 | 60 | 5144694 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC036559.1, NM_001159401.1 transcripts.
Set Membership: |
|

Upp1
Genomic Map