Assay Details
| Gene Symbol: Upp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159401.1 | NP_001152873.1 | 3 - 4 | 300 | 81 | |
| NM_001159402.1 | NP_001152874.1 | 3 - 4 | 299 | 81 | ||
| NM_009477.3 | NP_033503.2 | 3 - 4 | 327 | 81 | ||
| GenBank mRNA | AK136474.1 | - | 3 - 4 | 180 | 81 | |
| AL118479.1 | - | 3 - 4 | 300 | 81 | ||
| AV447018.1 | - | 3 - 4 | 351 | 81 | ||
| BC036559.1 | - | 3 - 4 | 201 | 5144694 | 81 | |
| CA493594.1 | - | 337 | 81 | |||
| CA536983.1 | - | 3 - 4 | 304 | 81 | ||
| D44464.1 | - | 3 - 4 | 328 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Upp1
Genomic Map