Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100001193.mm.1 |
NM_001159401.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001193.mm.1 |
NM_001159401.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001193.mm.1 |
NM_001159402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001193.mm.1 |
NM_001159402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001193.mm.1 |
NM_009477.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001193.mm.1 |
NM_009477.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001193.mm.1 |
TC1100000119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001191.mm.1 |
NM_001159402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001191.mm.1 |
NM_001159402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001191.mm.1 |
TC1100000119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001189.mm.1 |
NM_001159402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001189.mm.1 |
NM_001159402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001189.mm.1 |
TC1100000119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001188.mm.1 |
NM_001159402.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001188.mm.1 |
NM_001159402.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001188.mm.1 |
TC1100000119.mm.1 |
MTA TC |