Assay Details
| Gene Symbol: Upp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009477.3 | NP_033503.2 | 1 - 2 | 181 | 75 | |
| GenBank mRNA | CA536983.1 | - | 1 - 2 | 158 | 75 | |
| D44464.1 | - | 1 - 2 | 182 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D44464.1, NM_009477.3 transcripts.
Set Membership: |
|

Upp1
Genomic Map