Assay Details
| Gene Symbol: Hus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF076845.1 | - | 1 - 2 | 74 | 64 | |
| AK046152.1 | - | 1 - 2 | 84 | 64 | ||
| AK088403.1 | - | 1 - 2 | 84 | 64 | ||
| AK135922.1 | - | 1 - 2 | 64 | 64 | ||
| BB611267.1 | - | 1 - 2 | 140 | 64 | ||
| BC047460.1 | - | 1 - 2 | 91 | 64 | ||
| BC061249.1 | - | 1 - 2 | 110 | 30249148 | 64 | |
| Y16894.2 | - | 1 - 2 | 106 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hus1
Genomic Map