Assay Details
| Gene Symbol: Hus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF076845.1 | - | 2 - 3 | 195 | 126 | |
| AK046152.1 | - | 2 - 3 | 205 | 126 | ||
| AK051839.1 | - | 1 - 2 | 630 | 126 | ||
| AK088403.1 | - | 2 - 3 | 205 | 126 | ||
| AK135922.1 | - | 2 - 3 | 185 | 126 | ||
| BB611267.1 | - | 2 - 3 | 261 | 126 | ||
| BC047460.1 | - | 2 - 3 | 212 | 126 | ||
| BC061249.1 | - | 2 - 3 | 231 | 30249148 | 126 | |
| Y16894.2 | - | 2 - 3 | 227 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hus1
Genomic Map