Assay Details
| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009622.1 | NP_033752.1 | 10 - 11 | 2003 | 88 | |
| GenBank mRNA | AF053980.1 | - | 10 - 11 | 1596 | 88 | |
| AK220469.1 | - | 4 - 5 | 474 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcy1
Genomic Map