Assay Details
| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009622.1 | NP_033752.1 | 6 - 7 | 1410 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adcy1| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009622.1 | NP_033752.1 | 6 - 7 | 1410 | 66 | |
Set Membership: |
|