Assay Details
| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009622.1 | NP_033752.1 | 9 - 10 | 1912 | 71 | |
| GenBank mRNA | AK220469.1 | - | 3 - 4 | 383 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adcy1
Genomic Map