Assay Details
| Gene Symbol: Ikzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036833.1 | - | 3 - 4 | 415 | 79 | |
| AK077815.1 | - | 3 - 4 | 713 | 79 | ||
| AK154175.1 | - | 3 - 4 | 438 | 79 | ||
| AK220235.1 | - | 3 - 4 | 382 | 79 | ||
More Information
Set Membership: |
|

Ikzf1
Genomic Map