Assay Details
| Gene Symbol: Prorsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163454.2 | NP_001156926.1 | 1 - 2 | 124 | 89 | |
| GenBank mRNA | AK077280.1 | - | 1 - 2 | 122 | 89 | |
| AK085965.1 | - | 1 - 2 | 101 | 89 | ||
| BC033930.1 | - | 1 - 2 | 97 | 1397170 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077280.1, AK085965.1, BC033930.1 transcripts.
Set Membership: |
|

Prorsd1
Genomic Map