Assay Details
| Gene Symbol: Prorsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163454.2 | NP_001156926.1 | 2 - 3 | 197 | 154 | |
| GenBank mRNA | AK077280.1 | - | 2 - 3 | 195 | 154 | |
| AK085965.1 | - | 2 - 3 | 174 | 154 | ||
| BC033930.1 | - | 2 - 3 | 170 | 1397170 | 154 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077280.1, AK085965.1, BC033930.1 transcripts.
Set Membership: |
|

Prorsd1
Genomic Map