Assay Details
| Gene Symbol: Rtn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194053.3 | NP_918942.1 | 2 - 3 | 847 | 81 | |
| GenBank mRNA | AY102282.1 | - | 2 - 3 | 804 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Rtn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194053.3 | NP_918942.1 | 2 - 3 | 847 | 81 | |
| GenBank mRNA | AY102282.1 | - | 2 - 3 | 804 | 81 | |
Set Membership: |
|