Assay Details
| Gene Symbol: Zmiz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005867.1 | NP_001005867.1 | 2 - 3 | 282 | 92 | |
| NM_028601.3 | NP_082877.2 | 2 - 3 | 282 | 92 | ||
| GenBank mRNA | BB645605.1 | - | 3 - 4 | 432 | 92 | |
| BC060652.1 | - | 2 - 3 | 220 | 30354393 | 92 | |
| CK617573.1 | - | 3 - 4 | 381 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060652.1, NM_001005867.1, NM_028601.3 transcripts.
Set Membership: |
|

Zmiz2
Genomic Map