Assay Details
| Gene Symbol: Lck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162432.1 | NP_001155904.1 | 7 - 8 | 721 | 66 | |
| NM_001162433.1 | NP_001155905.1 | 7 - 8 | 763 | 66 | ||
| NM_010693.3 | NP_034823.1 | 6 - 7 | 818 | 66 | ||
| GenBank mRNA | AK088001.1 | - | 6 - 7 | 819 | 66 | |
| AK166263.1 | - | 1 - 2 | 208 | 66 | ||
| BC011474.1 | - | 7 - 8 | 728 | 4191610 | 66 | |
| CA577097.1 | - | 4 - 5 | 454 | 66 | ||
| M12056.1 | - | 6 - 7 | 794 | 66 | ||
| X03533.1 | - | 6 - 7 | 824 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lck
Genomic Map