Assay Details
| Gene Symbol: Ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016895.4 | NP_058591.2 | 6 - 7 | 811 | 66 | |
| GenBank mRNA | AK010936.1 | - | 6 - 7 | 730 | 66 | |
| AK010951.1 | - | 6 - 7 | 732 | 66 | ||
| AK077519.1 | - | 6 - 7 | 735 | 66 | ||
| AK166976.1 | - | 6 - 7 | 722 | 66 | ||
| AK167797.1 | - | 6 - 7 | 769 | 66 | ||
| AK168056.1 | - | 6 - 7 | 734 | 66 | ||
| AK168148.1 | - | 6 - 7 | 761 | 66 | ||
| BC008610.1 | - | 6 - 7 | 721 | 3584786 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ak2
Genomic Map