Assay Details
| Gene Symbol: Map7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198635.1 | NP_001185564.1 | 4 - 5 | 502 | 93 | |
| NM_008635.2 | NP_032661.2 | 4 - 5 | 502 | 93 | ||
| GenBank mRNA | AB098611.1 | - | 4 - 5 | 408 | 93 | |
| AK008018.1 | - | 4 - 5 | 446 | 93 | ||
| AK008182.1 | - | 4 - 5 | 425 | 93 | ||
| AK014575.1 | - | 4 - 5 | 419 | 93 | ||
| AK133267.1 | - | 4 - 5 | 951 | 93 | ||
| BC042771.1 | - | 4 - 5 | 427 | 93 | ||
| BC044733.1 | - | 4 - 5 | 427 | 93 | ||
| BC046555.1 | - | 4 - 5 | 427 | 93 | ||
| BC052637.1 | - | 4 - 5 | 438 | 6467153 | 93 | |
| Y15197.1 | - | 4 - 5 | 408 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map7
Genomic Map